missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-methylhepta-1,6-diene, TRC

Product Code. 30400140
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400140 2.5 g 2.5g
30408728 5 g -
30391963 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400140 Supplier Toronto Research Chemicals Supplier No. TRCH9474252.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25201-40-5
Molecular Formula C8 H14 O
Molecular Weight (g/mol) 126.2
IUPAC Name 4-methylhepta-1,6-dien-4-ol
SMILES CC(O)(CC=C)CC=C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H14O/c1-4-6-8(3,9)7-5-2/h4-5,9H,1-2,6-7H2,3H3
Formula Weight 126.1045
Chemical Name or Material 4-Hydroxy-4-methylhepta-1,6-diene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.