missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4-methyl-2-pentanone, 98+%

Product Code. 11467707
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467707 500 mL 500mL
11334719 100 mL 100mL
11477707 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467707 Supplier Thermo Scientific Alfa Aesar Supplier No. A16248.AP
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 123-42-2 | C6H12O2 | 116.16 g/mol

4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatings…). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatings…). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-42-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004471
InChI Key SWXVUIWOUIDPGS-UHFFFAOYSA-N
Synonym diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
PubChem CID 31256
ChEBI CHEBI:55381
IUPAC Name 4-hydroxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)O

Specifications

Melting Point -47°C
Density 0.935
Boiling Point 168°C
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.423
Quantity 500 mL
UN Number UN1148
Beilstein 1740440
Merck Index 14,2964
Solubility Information It is soluble in water.
Formula Weight 116.16
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-4-methyl-2-pentanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.