missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-4'-iodobiphenyl, 98+%

Product Code. 11469627
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469627 5 g 5g
11479627 25 g 25g
11489627 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469627 Supplier Thermo Scientific Alfa Aesar Supplier No. B22115.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 29558-78-9 | C12H9IO | 296.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29558-78-9
Molecular Formula C12H9IO
Molecular Weight (g/mol) 296.107
MDL Number MFCD02258843
InChI Key WJXIAMCDWSUSEI-UHFFFAOYSA-N
Synonym 4-hydroxy-4'-iodobiphenyl, 4'-iodo-1,1'-biphenyl-4-ol, 4-4-iodophenyl phenol, 4-4'-iodophenyl phenol, 4'-iodobiphenyl-4-ol, acmc-20amgy, pubchem9061, 4-4?iodophenyl phenol, 4-4'-iodophenyl-phenol
PubChem CID 2064030
IUPAC Name 4-(4-iodophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)O

Specifications

Melting Point 196°C to 202°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 296.11
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-4'-iodobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.