missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%

Product Code. 15424247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424247 1 g 1g
15434247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424247 Supplier Thermo Scientific Alfa Aesar Supplier No. H31639.03
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124811-71-8 | C8H4F3NO | 187.121 g/mol

As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124811-71-8
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD06797882
InChI Key QXIFYONUKBXFTH-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile
PubChem CID 2783159
IUPAC Name 4-hydroxy-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)C(F)(F)F)O

Specifications

Melting Point 171°C to 175°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 187.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.