missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11302069
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302069 1 g 1g
15467545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302069 Supplier Thermo Scientific Alfa Aesar Supplier No. H26097.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220239-68-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD01091008
InChI Key DPVRVZQEDJVWLS-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzoicacid, acmc-1cdpd, ksc495o6p, 4-hydroxy-3-trifluoromethyl-benzoic acid, 4-oxidanyl-3-trifluoromethyl benzoic acid
PubChem CID 2775112
IUPAC Name 4-hydroxy-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F

Specifications

Melting Point 144°C to 147°C
Quantity 1 g
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.