missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
Melting Point | 144.0°C to 147.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | CF3C6H3CO2H(OH) |
Quantity | 1 g |
Formula Weight | 206.12 |
Percent Purity | 97% |
Chemical Name or Material | 4-Hydroxy-3-(trifluoromethyl)benzoic acid |
Safety and Handling
Product Identifier
- 4-Hydroxy-3-(trifluoromethyl)benzoic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'