missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 15405357
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405357 1 g 1g
15468135 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405357 Supplier Thermo Scientific Alfa Aesar Supplier No. H32602.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53104-95-3 | Molecular Formula: C8H5F3O3 | Molecular Weight: 206.12 g/mol

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53104-95-3
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD04973780
InChI Key GPJSLRQIOKFRFL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde
PubChem CID 7018050
IUPAC Name 4-hydroxy-3-(trifluoromethoxy)benzaldehyde
SMILES OC1=C(OC(F)(F)F)C=C(C=O)C=C1

Specifications

Melting Point 76°C to 80°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.