missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(p-diazophenylphosphorylcholine) Phenylacetic Acid, TRC

Product Code. 30396316
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
missing translation for 'unitSize'
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396316 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30396316 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCH94100025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-3-(p-diazophenylphosphorylcholine) Phenylacetic Ac
CAS 359435-74-8
Molecular Formula C19 H24 N3 O7 P
Quantity 25 mg
Synonym Ethanaminium, 2-[[[4-[2-[5-(carboxymethyl)-2-hydroxyphenyl]diazenyl]phenoxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt, Ethanaminium, 2-[[[4-[[5-(carboxymethyl)-2-hydroxyphenyl]azo]phenoxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt (9CI), 4-Hydroxy-3-(p-diazophenylphosphorylcholine)phenylacetic acid
InChI Formula InChI=1S/C19H24N3O7P/c1-22(2,3)10-11-28-30(26,27)29-16-7-5-15(6-8-16)20-21-17-12-14(13-19(24)25)4-9-18(17)23/h4-9,12H,10-11,13H2,1-3H3,(H2-,20,23,24,25,26,27)
SMILES C[N+](C)(C)CCOP(=O)([O-])Oc1ccc(cc1)N=Nc2cc(CC(=O)O)ccc2O
IUPAC Name [4-[(E)-[5-(carboxymethyl)-2-hydroxyphenyl]diazenyl]phenyl] 2-(trimethylazaniumyl)ethyl phosphate
Molecular Weight (g/mol) 437.38
Formula Weight 437.1352
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.