missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%, ACROS Organics™

Brand:  Acros Organics 204890250

Code : NEW

Additional Details : CAS Number : 10463-20-4 Weight : 0.03000kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 13428458

  • £187.20 / 25g

Estimated Shipment date
from Supplier 22-04-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

10463-20-4
197.15
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
CHEBI:546274
C8H7NO5
QBHBHOSRLDPIHG-UHFFFAOYSA-N
447364
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O

Specifications

4-Hydroxy-3-nitrophenylacetic acid
Glass bottle
144.0°C to 147.0°C
99%
98.5
98.5% min. (HPLC)
HOC6H3(NO2)CH2CO2H
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O
447364
197.15
99%
Authentic
Colorless to Yellow
25g
10463-20-4
100.0
C8H7NO5
10,IV,562
QBHBHOSRLDPIHG-UHFFFAOYSA-N
197.15
CHEBI:546274
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxy-3-nitrophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents