missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methylbenzoic acid hydrate, 97%, Thermo Scientific™

Product Code. 11322989
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11322989

Brand: Thermo Scientific Alfa Aesar H27792.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00270105
InChI Key LTFHNKUKQYVHDX-UHFFFAOYSA-N
Synonym 4-hydroxy-m-toluic acid, 3-methyl-4-hydroxybenzoic acid, 4-hydroxy-3-methyl-benzoic acid, 4,3-cresotic acid, 4-hydroxy-3-methylbenzoate, benzoic acid, 4-hydroxy-3-methyl, 4-hydroxy-m-toluate, pubchem20044, acmc-209kj4, 3-methyl-4-hydroxybenzoate
PubChem CID 68138
ChEBI CHEBI:85239
IUPAC Name 4-hydroxy-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1O)C(O)=O

Specifications

Melting Point 173°C to 177°C
Odor Odorless
Quantity 1 g
Formula Weight 152.15 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3-methylbenzoic acid hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.