missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methyl-2-butanone, tech 85%

Product Code. p-7030409
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11412060 1 g 1g
11422060 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11412060

Brand: Thermo Scientific Alfa Aesar H28483.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3393-64-4 | C5H10O2 | 102.133 g/mol

Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo vinylation and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo “vinylation” and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3393-64-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004739
InChI Key VVSRECWZBBJOTG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl, 3-hydroxymethyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl-, +, acmc-1cngd, 3-methyl-4-hydroxy-2-butanone, vvsrecwzbbjotg-uhfffaoysa, 2-butanone,4-hydroxy-3-methyl, 4-hydroxy-3-methyl-2-butanone, technical grade
PubChem CID 18829
IUPAC Name 4-hydroxy-3-methylbutan-2-one
SMILES CC(CO)C(=O)C

Specifications

Refractive Index 1.4325
Quantity 1 g
Formula Weight 102.13
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Hydroxy-3-methyl-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.