missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxymandelic Acid, TRC

Product Code. 30003434
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003434 250 mg 250mg
17904055 100 mg 100mg
17903865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003434 Supplier Toronto Research Chemicals Supplier No. TRCH946330250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9 H10 O5
Molecular Weight (g/mol) 198.17
Synonym 4-Hydroxy-3-methoxymandelic Acid, Vanillylmandelic acid, Benzeneacetic acid, α,4-dihydroxy-3-methoxy- (9CI, ACI), Mandelic acid, 4-hydroxy-3-methoxy- (8CI), α,4-Dihydroxy-3-methoxybenzeneacetic acid (ACI), (4-Hydroxy-3-methoxyphenyl)glycolic acid, (±)-3-Methoxy-4-hydroxymandelic acid, (±)-4-Hydroxy-3-methoxymandelic acid, (±)-Vanilmandelic acid, 2-(4'-Hydroxy-3'-methoxyphenyl)-2-hydroxyethanoic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, 3-Methoxy-4-hydroxyphenylhydroxyacetic acid, 4-Hydroxy-3-methoxy-DL-mandelic acid, 4-Hydroxy-3-methoxymandelic acid, 4'-Hydroxy-3'-methoxymandelic acid, DL-3-Methoxy-4-hydroxymandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, DL-Vanillomandelic acid, DL-Vanillylmandelic acid, dl-4-Hydroxy-3-methoxymandelic acid (dl-vanillylmandelic acid), dl-Vanillomandelic acid, HMMA, MeSH ID: D014642, Vanillinmandelic acid, Vanillomandelic acid, Vanillyl-dl-mandelic acid, Vanilmandelic acid, VMA
IUPAC Name 2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COc1cc(ccc1O)C(O)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)
Formula Weight 198.0528
Chemical Name or Material 4-Hydroxy-3-methoxymandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.