missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxymandelic Acid-13C6, TRC

Product Code. 30395916
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395916 1 mg -
30401654 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395916 Supplier Toronto Research Chemicals Supplier No. TRCH9463331MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189690-96-7
Molecular Formula C313C6H10O5
Molecular Weight (g/mol) 204.13
Synonym α,4-Dihydroxy-3-methoxybenzeneacetic Acid-13C6, (+/-)-Vanilmandelic Acid-13C6, (4-Hydroxy-3-methoxyphenyl)glycolic Acid-13C6, 3-Methoxy-4-hydroxyphenyl- hydroxyacetic Acid-13C6, HMMA-13C6, VMA-13C6, Vanilinmandelic Acid-13C6, dl-Vanillomandelic Acid-13C6
IUPAC Name 2-hydroxy-2-(4-hydroxy-3-methoxy(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl)acetic acid
SMILES O[13C]1=[13C](OC)[13CH]=[13C](C(O)C(O)=O)[13CH]=[13CH]1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)/i2+1,3+1,4+1,5+1,6+1,7+1
Formula Weight 204.073
Chemical Name or Material 4-Hydroxy-3-methoxymandelic Acid-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.