missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%

Product Code. 10464745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10464745 5 g Glass bottle 5g
10150122 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10464745 Supplier Thermo Scientific Acros Supplier No. 121370050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7149-10-2 | C8H11NO2·HCl | 189.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-2-7149
Molecular Formula C8H11NO2·HCl
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012864
InChI Key LOYPVODLNGWOLM-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methoxybenzylamine hydrochloride, 4-aminomethyl-2-methoxyphenol hydrochloride, vanillylamine hydrochloride, n-vanillylamine hydrochloride, 2-vanillylamine hydrochloride, 4-hydroxy-3-methoxybenzylamine hcl, phenol, 4-aminomethyl-2-methoxy-, hydrochloride, 4-aminomethyl-2-methoxyphenol, chloride, vanillylamine hcl
PubChem CID 165576
IUPAC Name 4-(aminomethyl)-2-methoxyphenol;hydrochloride
SMILES COC1=C(C=CC(=C1)CN)O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 219°C to 221°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CH2NH2·HCl
Quantity 5 g
Beilstein 13, I, 321
Formula Weight 189.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-3-methoxybenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.