Learn More
4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Description
4'-Hydroxy-3'-methoxyacetophenone is used as an inhibitor of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase, which is an enzyme responsible for reactive oxygen species production. It is also useful in the treatment of various inflammatory diseases and atherosclerosis. Further, it shows an anti-inflammatory activity and improves endothelial function by reducing oxidative stress. In addition to this, it is used for studying nitric oxide regulated processes in plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in ethanol, chloroform, ether, hot water, benzene, ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 112°C to 116°C |
Boiling Point | 295°C to 300°C |
Quantity | 250 g |
Beilstein | 637373 |
Merck Index | 14,741 |
Solubility Information | Soluble in ethanol,chloroform,ether,hot water,benzene,ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether. |
Formula Weight | 166.18 |
Percent Purity | 98% |
Chemical Name or Material | 4'-Hydroxy-3'-methoxyacetophenone |
RUO – Research Use Only