missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-isoxazoledicarboxylic Acid 3,5-Diethyl Ester, TRC

Product Code. 30391671
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391671 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391671 Supplier Toronto Research Chemicals Supplier No. TRCH94361010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-3,5-isoxazoledicarboxylic Acid 3,5-Diethyl Ester
CAS 23468-99-7
Molecular Formula C9H11NO6
Quantity 10 mg
Synonym 4-Hydroxy-3,5-isoxazoledicarboxylic Acid Diethyl Ester
InChI Formula InChI=1S/C9H11NO6/c1-3-14-8(12)5-6(11)7(16-10-5)9(13)15-4-2/h11H,3-4H2,1-2H3
SMILES CCOC(=O)c1onc(C(=O)OCC)c1O
IUPAC Name diethyl 4-hydroxy-1,2-oxazole-3,5-dicarboxylate
Molecular Weight (g/mol) 229.19
Formula Weight 229.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.