missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzonitrile, 97%, Thermo Scientific™

Product Code. p-4510338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10633075 1 g 1g
10311665 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10633075

Brand: Thermo Scientific Maybridge RDP00227DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4198-90-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
InChI Key WFYGXOWFEIOHCZ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm
PubChem CID 20176
IUPAC Name 4-hydroxy-3,5-dimethylbenzonitrile
SMILES CC1=CC(=CC(=C1O)C)C#N

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 147.18
Percent Purity 95%
Chemical Name or Material 4-Hydroxy-3, 5-dimethylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.