missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%

Product Code. 11372229
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372229 1 g 1g
11382229 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372229 Supplier Thermo Scientific Alfa Aesar Supplier No. H27231.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4385-56-2 | C10H12O5 | 212.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4385-56-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00016834
InChI Key BQBQKSSTFGCRQL-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-4-hydroxyphenylacetic acid, homosyringic acid, 2-4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenylacetic acid, benzeneacetic acid, 4-hydroxy-3,5-dimethoxy, acmc-1aewn, benzeneacetic acid,4-hydroxy-3,5-dimethoxy, 4-hydroxy-3,5-dimethoxy-phenyl-acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid #
PubChem CID 78093
IUPAC Name 2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1O)OC)CC(=O)O

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Beilstein 2851658
Formula Weight 212.21
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.