missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxybenzyl Alcohol, TRC

Product Code. 30396225
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396225 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396225 Supplier Toronto Research Chemicals Supplier No. TRCH972693500MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzyl Alcohol
CAS 530-56-3
Molecular Formula C9 H12 O4
Quantity 500 mg
Synonym 4-Hydroxy-3,5-dimethoxybenzenemethanol, 2,6-Dimethoxy-4-(hydroxymethyl)phenol, 3,5-Dimethoxy-4-hydroxybenzyl Alcohol, 4-Hydroxy-3,5-dimethoxybenzyl Alcohol, Syringic Alcohol, Syringyl Alcohol
InChI Formula InChI=1S/C9H12O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-4,10-11H,5H2,1-2H3
SMILES COc1cc(CO)cc(OC)c1O
IUPAC Name 4-(hydroxymethyl)-2,6-dimethoxyphenol
Molecular Weight (g/mol) 184.189
Formula Weight 184.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.