missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%

Product Code. 11413960
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413960 1 g 1g
11451588 5 g 5g
11461588 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413960 Supplier Thermo Scientific Alfa Aesar Supplier No. A13888.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 530-56-3 | C9H12O4 | 184.191 g/mol

4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

Solubility
Sparingly soluble (0.40 g/L at 25°C).

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-56-3
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD00016871
InChI Key LUOAEJWSKPQLJD-UHFFFAOYSA-N
Synonym 4-hydroxy-3,5-dimethoxybenzyl alcohol, 4-hydroxymethyl-2,6-dimethoxyphenol, syringyl alcohol, syringic alcohol, 3,5-dimethoxy-4-hydroxybenzyl alcohol, unii-k7g66i59xc, benzenemethanol, 4-hydroxy-3,5-dimethoxy, 2,6-dimethoxy-4-hydroxymethyl phenol, 4-hydroxymethyl-2,6-dimethoxy-phenol, 3,5-dimethoxy-4-hydroxybenzylalcohol
PubChem CID 10741
ChEBI CHEBI:86948
IUPAC Name 4-(hydroxymethyl)-2,6-dimethoxyphenol
SMILES COC1=CC(=CC(=C1O)OC)CO

Specifications

Melting Point 129°C to 132°C
Quantity 1 g
Beilstein 1873975
Solubility Information Sparingly soluble (0.40g/L at 25°C).
Formula Weight 184.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.