missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzaldehyde, 98+%

Product Code. 11374434
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374434 50 g 50g
11364434 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374434 Supplier Thermo Scientific Alfa Aesar Supplier No. L01060.18
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1948-40-9 | C7H4I2O2 | 373.92 g/mol

4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-diiodobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Sparingly soluble in chloroform and methanol.

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1948-40-9
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.92
MDL Number MFCD00014670
InChI Key WHLUEIMENHLCMY-UHFFFAOYSA-N
Synonym 3,5-diiodo-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-diiodo, 2,6-diiodo-4-formylphenol, 4-hydroxy-3,5-diiodo-benzaldehyde, acmc-1c01x, #, benzaldehyde,4-hydroxy-3,5-diiodo
PubChem CID 74760
IUPAC Name 4-hydroxy-3,5-diiodobenzaldehyde
SMILES OC1=C(I)C=C(C=O)C=C1I

Specifications

Melting Point 198°C to 206°C
Quantity 50 g
Beilstein 1283414
Sensitivity Air and light sensitive
Solubility Information Insoluble in water. Sparingly soluble in chloroform and methanol.
Formula Weight 373.92
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3,5-diiodobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.