missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2H-1-benzothiopyran-2-one, TRC

Product Code. 30390379
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390379 100 mg -
30393450 500 mg 500mg
30406424 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390379 Supplier Toronto Research Chemicals Supplier No. TRCH808493100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16854-67-4
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
Synonym 4-Hydroxy-1-(2H)-benzothiopyran-2-one, 4-Hydroxy-1-thiacoumarin, 4-Hydroxy-1-thiocoumarin, 4-Hydroxy-2-oxo-2H-1-benzothiopyran, 4-Hydroxythiocoumarin
IUPAC Name 4-hydroxythiochromen-2-one
SMILES OC1=CC(=O)Sc2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H6O2S/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5,10H
Formula Weight 178.0089
Chemical Name or Material 4-Hydroxy-2H-1-benzothiopyran-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.