missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-2'-methylacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 10147903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10147903 5 g Glass bottle 5g
10588470 25 g Glass bottle 25g
10331624 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147903 Supplier Thermo Scientific Acros Supplier No. 121390050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 875-59-2 | Molecular Formula: C9H10O2 | Molecular Weight: 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-59-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002303
InChI Key IAMNVCJECQWBLZ-UHFFFAOYSA-N
PubChem CID 70133
ChEBI CHEBI:87314
IUPAC Name 1-(4-hydroxy-2-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 127°C to 132°C
Color Tan to White
Boiling Point 313°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(CH3)COCH3
Quantity 5 g
Beilstein 08, 111
Synonym 4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 150.18
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 4'-Hydroxy-2'-methylacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.