missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methyl-2H-thieno-[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide, Mikromol™

Product Code. 17942076
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942076 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17942076 Supplier Mikromol Supplier No. MM0247.08
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Hydroxy-2-methyl-2H-thieno-[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-2-methyl-2H-thieno-[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide
CAS 105410-48-8
Analyte or Component Names 4-Hydroxy-2-methyl-2H-thieno-[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide
Shipping Condition Room Temperature
Molecular Formula C8 H8 N2 O4 S2
Certifications/Compliance ISO 17025
Synonym Tenoxicam Imp. G (EP), 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide, Tenoxicam Impurity G
InChI Formula InChI=1S/C8H8N2O4S2/c1-10-5(8(9)12)6(11)7-4(2-3-15-7)16(10,13)14/h2-3,11H,1H3,(H2,9,12)
SMILES CN1C(=C(O)c2sccc2S1(=O)=O)C(=O)N
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxothieno[2,3-e]thiazine-3-carboxamide
Molecular Weight (g/mol) 260.29
Quantity 50 mg
Formula Weight 259.9925 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.