missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methyl- 1,1-dioxide-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide, TRC

Product Code. 30404409
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404409 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404409 Supplier Toronto Research Chemicals Supplier No. TRCH620485100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-2-methyl-2H-thieno-[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide
CAS 105410-48-8
Molecular Formula C8 H8 N2 O4 S2
Quantity 100 mg
Synonym Tenoxicam Imp. G (EP), 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxamide 1,1-Dioxide
InChI Formula InChI=1S/C8H8N2O4S2/c1-10-5(8(9)12)6(11)7-4(2-3-15-7)16(10,13)14/h2-3,11H,1H3,(H2,9,12)
SMILES CN1C(=C(O)c2sccc2S1(=O)=O)C(=O)N
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxothieno[2,3-e]thiazine-3-carboxamide
Molecular Weight (g/mol) 260.29
Formula Weight 259.9925
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.