missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-2-hexenal Antibody (6F10), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59352PEATT594
This item is not returnable.
View return policy
Description
4-Hydroxy-2-hexenal Monoclonal specifically detects 4-Hydroxy-2-hexenal in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
| 4-Hydroxy-2-hexenal | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Specific for 4-Hydroxy-2-hexenal (4-HHE) and 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxy-2-hexenal (4-HHE) or 4-Hydroxynonenal (4-HNE). Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 6F10 | |
| Flow Cytometry | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction