missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-hexenal Antibody (5C11.1), Novus Biologicals™

Product Code. 18327936 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327936 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327936 Supplier Novus Biologicals Supplier No. NBP318263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in All species samples. It is validated for Western Blot, ELISA, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Hydroxy-2-hexenal
Applications Western Blot, ELISA, Dot Blot
Classification Monoclonal
Clone 5C11.1
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000, Dot Blot
Formulation PBS pH 7.4, 50% glycerol
Host Species Mouse
Immunogen Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH)
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.