missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2(5H)-furanone, 95%

Product Code. 10209743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209743 5 g Glass bottle 5g
10216290 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209743 Supplier Thermo Scientific Acros Supplier No. 235260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-57-1 | C4H4O3 | 100.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-57-1
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00052187
InChI Key JZQBAGOECGRTSA-UHFFFAOYSA-N
Synonym 4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g
PubChem CID 54683813
IUPAC Name 3-hydroxy-2H-furan-5-one
SMILES OC1=CC(=O)OC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 135°C to 137°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,acetonitrile,dmso
Formula Weight 100.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2(5H)-furanone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-2(5H)-furanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hydroxyfuran-2(5H)-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.