missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical

Product Code. 10144480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144480 5 g Glass bottle 5g
10337740 1 g Glass bottle 1g
10399813 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10144480 Supplier Thermo Scientific Acros Supplier No. 175050050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2226-96-2 | C9H18NO2 | 172.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9H18NO2
Molecular Weight (g/mol) 172.25
MDL Number MFCD00006478
Synonym 4-Hydroxy-TEMPO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 73.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Fieser 09,247
Merck Index 15, 9284
Solubility Information Solubility in water: soluble.
Viscosity 20 mPa.s (40°C)
Formula Weight 172.25
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, Free radical
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.