missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyloxy, TRC

Product Code. 30393356
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393356 5 g -
30406450 10 g 10g
30406198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393356 Supplier Toronto Research Chemicals Supplier No. TRCH9565005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9 H18 N O2
Molecular Weight (g/mol) 172.24
Synonym 1-Piperidinyloxy, 4-hydroxy-2,2,6,6-tetramethyl- (9CI, ACI), 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyloxy (ACI), Piperidinooxy, 4-hydroxy-2,2,6,6-tetramethyl- (7CI, 8CI), 1-Oxyl-2,2,6,6-tetramethyl-4-hydroxypiperidine, 1-Oxyl-2,2,6,6-tetramethyl-4-piperidinol, 2,2,6,6-Tetramethyl-1-oxy-4-hydroxypiperidine, 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 2,2,6,6-Tetramethyl-4-hydroxylpiperidine-1-oxyl, 2,2,6,6-Tetramethyl-4-hydroxypiperidin-1-oxyl, 2,2,6,6-Tetramethyl-4-hydroxypiperidine 1-oxide radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidine N-oxide, 2,2,6,6-Tetramethyl-4-hydroxypiperidine oxide, 2,2,6,6-Tetramethyl-4-hydroxypiperidine-1-hydroxyl, 2,2,6,6-Tetramethyl-4-hydroxypiperidine-1-oxyl, 2,2,6,6-Tetramethyl-4-hydroxypiperidine-1-oxyl radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidinooxy, 2,2,6,6-Tetramethyl-4-hydroxypiperidinooxy radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidinyl-1-oxy, 2,2,6,6-Tetramethyl-4-hydroxypiperidinyl-1-oxyl, 2,2,6,6-Tetramethyl-4-hydroxypiperidinyloxy radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidinyloxyl radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidyl 1-oxyl, 2,2,6,6-Tetramethyl-4-oxypiperidine-1-oxyl, 2,2,6,6-Tetramethyl-4-piperidinol 1-oxide, 2,2,6,6-Tetramethyl-4-piperidinol 1-oxyl, 2,2,6,6-Tetramethyl-4-piperidinol N-oxyl, 2,2,6,6-Tetramethyl-4-piperidinol nitroxide, 2,2,6,6-Tetramethyl-4-piperidinol-1-oxy, 2,2,6,6-Tetramethyl-4-piperidinol-1-oxyl radical, 2,2,6,6-Tetramethylpiperidine-4-hydroxy-1-oxyl, 2,2,6,6-Tetramethylpiperidine-N-oxyl-4-ol, 2,2,6,6-Tetramethylpiperidinol-4-oxyl-1, 4-HO-TEMPO, 4-Hydroxy-1-oxyl-2,2,6,6-tetramethylpiperidine, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinoxy, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinoxyl, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxide radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine N-oxide, 4-Hydroxy-2,2,6,6-tetramethylpiperidine oxide, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-N-oxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-N-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidino-1-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidinoxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidinoxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyl-1-oxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyl-1-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyl-N-oxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyl-N-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyloxy, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyloxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidyl-1-oxyl, 4-hydroxy-TEMPO, 4-OH-Tempo, 4-Oxypiperidol, 4H-Tempo, 701, Adeka Stab 7RD, ADK Stab LA 7RD, ADK Stab NA 7RD, H 0865, HOTEMPO, HTEMPO, HyTEMPO, LA 7RD, Lignostab 1198L, Nitroxyl 2, NR I (4-Hydroxy-TEMPO), Petroflo 20Y104, Polymerization inhibitor 701, Polystast 7200P, Polystop 7010, Polystop 7200P, Polystop 7300P, Prostab 5198, SPJ 701, Tanol, Tempo OH, Tempol, Tempole, Tetramethyl-2,2,6,6-aza-1-cyclohexanol-4-oxide-1, TMPN, ZJ 01, ZJ 701
IUPAC Name 1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-ol
SMILES CC1(C)CC(O)CC(C)(C)N1[O]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11H,5-6H2,1-4H3
Formula Weight 172.1338
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyloxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.