missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-1H-benzotriazole, 98%

Product Code. 11405580
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405580 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405580 Supplier Alfa Aesar™ Supplier No. H25843.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26725-51-9
Molecular Formula C6H5N3O
MDL Number MFCD00970892
Quantity 1g
Synonym 4-hydroxybenzotriazole, 4-hydroxy-1h-benzotriazole, hydroxybenzotriazole, 1h-1,2,3-benzotriazol-4-ol, hydroxybenztriazole, 1h-benzo d 1,2,3 triazol-4-ol, 1h-benzotriazol-4-ol, 3h-1,2,3-benzotriazol-4-ol, hydroxybenztriazol, hydroxybezotriazol
InChI Key NPZTUJOABDZTLV-UHFFFAOYSA-N
SMILES C1=CC(=O)C2=NNNC2=C1
IUPAC Name 1,2-dihydrobenzotriazol-4-one
Molecular Weight (g/mol) 135.126
PubChem CID 2759291
Formula Weight 135.13
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-1H-benzotriazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.