missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine, 98+%, Alfa Aesar™

Product Code. 11358345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358345 5 g 5g
11348345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358345 Supplier Thermo Scientific Alfa Aesar Supplier No. L10286.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 21314-17-0
Molecular Formula C11H8N4O
Molecular Weight (g/mol) 212.21
MDL Number MFCD00082637
InChI Key SVKPVSOBZKPDIY-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ol, 4-hydroxy-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one, 4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5-dihydro-1-phenyl, 1-phenyl-2h-pyrazolo 3,4-d pyrimidin-4-one, gdzqghgckkwnac-uhfffaoysa-n, 1-phenylpyrazolo 3,4-d pyrimidin-4-ol, 1-phenyl-5h-pyrazolo 3,4-d pyrimidin-4-one, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4 5h-one, 1-phenyl-5-hydropyrazolo 5,4-d pyrimidin-4-one
PubChem CID 88867
IUPAC Name 1-phenyl-2H-pyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2N(NC=C12)C1=CC=CC=C1

Specifications

Melting Point 304°C to 307°C
Quantity 5 g
Beilstein 198880
Formula Weight 212.21
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.