missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-methylpiperidine, 98%

Product Code. 11332516
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332516 500 g 500g
11322516 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332516 Supplier Thermo Scientific Alfa Aesar Supplier No. L06415.36
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-52-5 | C6H13NO | 115.176 g/mol

4-Hydroxy-1-methylpiperidine is used as reactant for optimization of Novobiocin scaffold to product antitumor agents, substitution about the rigidifying ring for histamine H4 receptor antagonist synthesis. It is also used as a reactant in the synthesis of CaMKII, VEGFR and FGFR kinase inhibitors and Phosphoinositide-3-kinase, Protein lysine methyltransferase G9a inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-1-methylpiperidine is used as reactant for optimization of Novobiocin scaffold to product antitumor agents, substitution about the rigidifying ring for histamine H4 receptor antagonist synthesis. It is also used as a reactant in the synthesis of CaMKII, VEGFR and FGFR kinase inhibitors and Phosphoinositide-3-kinase, Protein lysine methyltransferase G9a inhibitors.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture and oxidizing agents. Incompatible with water and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-52-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00006500
InChI Key BAUWRHPMUVYFOD-UHFFFAOYSA-N
Synonym 4-hydroxy-1-methylpiperidine, n-methyl-4-piperidinol, 1-methyl-4-piperidinol, 4-hydroxy-n-methylpiperidine, n-methyl-4-hydroxypiperidine, 4-piperidinol, 1-methyl, 1-methyl-4-hydroxypiperidine, n-methylpiperidol, n-methyl-4-hydroxy piperidine, 4-hydroxy-1-methyl-piperidine
PubChem CID 66048
IUPAC Name 1-methylpiperidin-4-ol
SMILES CN1CCC(CC1)O

Specifications

Melting Point 26°C to 30°C
Density 0.98
Boiling Point 198°C to 200°C
Flash Point 87°C (188°F)
Refractive Index 1.479
Quantity 500 g
Beilstein 103024
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-1-methylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.