missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-indanone, 97%, Thermo Scientific™

Product Code. 15482618
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482618 1 g 1g
15492618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482618 Supplier Thermo Scientific Alfa Aesar Supplier No. H56184.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Hydroxy-1-indanone is a hydroxylated indanone used for synthesis. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in ethanol. (25mg/ml)

Notes
Incompatible with oxidizing agents. Store at room temperature. Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40731-98-4
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00143330
InChI Key CKSCMRNFDBWFND-UHFFFAOYSA-N
Synonym 4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
PubChem CID 590547
IUPAC Name 4-hydroxy-2,3-dihydroinden-1-one
SMILES OC1=CC=CC2=C1CCC2=O

Specifications

Melting Point 242°C to 244°C
Quantity 1 g
Solubility Information Soluble in ethanol. (25mg/ml)
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.