missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroperoxy Cyclophosphamide, TRC

Product Code. 30006882
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006882 10 mg 10mg
30008246 5 mg 5mg
30010507 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006882 Supplier Toronto Research Chemicals Supplier No. TRCH71467510MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39800-16-3
Molecular Formula C7 H15 Cl2 N2 O4 P
Molecular Weight (g/mol) 293.08
IUPAC Name N,N-bis(2-chloroethyl)-4-hydroperoxy-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine
SMILES OOC1CCOP(=O)(N1)N(CCCl)CCCl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H15Cl2N2O4P/c8-2-4-11(5-3-9)16(13)10-7(15-12)1-6-14-16/h7,12H,1-6H2,(H,10,13)
Formula Weight 292.0146
Chemical Name or Material 4-Hydroperoxy Cyclophosphamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.