missing translation for 'onlineSavingsMsg'
Learn More

4-HO-DPHP, TRC

Product Code. 30396685
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396685 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396685 Supplier Toronto Research Chemicals Supplier No. TRCH4564205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-HO-DPHP
CAS 114527-61-6
Purity Grade Notes HPLC
Molecular Formula C12 H11 O5 P
Quantity 5 mg
Synonym 4-Hydroxyphenyl Phenyl Hydrogen Phosphate, 4-Hydroxyphenyl Diphenyl Phosphate, Phosphoric Acid (4-Hydroxyphenyl)diphenyl Ester
InChI Formula InChI=1S/C12H11O5P/c13-10-6-8-12(9-7-10)17-18(14,15)16-11-4-2-1-3-5-11/h1-9,13H,(H,14,15)
SMILES Oc1ccc(OP(=O)(O)Oc2ccccc2)cc1
IUPAC Name (4-hydroxyphenyl) phenyl hydrogen phosphate
Molecular Weight (g/mol) 266.187
Formula Weight 266.034
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.