missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylpyridine, TRC

Product Code. 30395638
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395638 50 mg -
30403383 100 mg 100mg
30399151 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395638 Supplier Toronto Research Chemicals Supplier No. TRCH32496050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27876-24-0
Molecular Formula C11 H17 N
Molecular Weight (g/mol) 163.259
IUPAC Name 4-hexylpyridine
SMILES CCCCCCc1ccncc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H17N/c1-2-3-4-5-6-11-7-9-12-10-8-11/h7-10H,2-6H2,1H3
Formula Weight 163.136
Chemical Name or Material 4-Hexylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.