missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10163444
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163444 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163444 Supplier Thermo Scientific Maybridge Supplier No. BTB09862DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 21643-38-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H18O2
Linear Formula CH3(CH2)4CH2C6H4COOH
MDL Number MFCD00013997
Quantity 1 g
Synonym benzoic acid, 4-hexyl, 4-n-hexylbenzoic acid, p-hexylbenzoic acid, benzoic acid, p-hexyl, p-n-hexylbenzoic acid, 4n-hexylbenzoic acid, maybridge1_002028, para-n-hexyl benzoic acid, acmc-1chu5, ksc201s3p
InChI Key CPEPWESLFZVUEP-UHFFFAOYSA-N
SMILES CCCCCCC1=CC=C(C=C1)C(=O)O
IUPAC Name 4-hexylbenzoic acid
Molecular Weight (g/mol) 206.285
PubChem CID 30725
Formula Weight 206.28
Percent Purity 97%
Chemical Name or Material 4-Hexylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hexylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.