missing translation for 'onlineSavingsMsg'
Learn More

4-Hexen-3-one, 98%, trans-isomer >95%

Product Code. 11307299
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307299 10 g 10g
11317299 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307299 Supplier Thermo Scientific Alfa Aesar Supplier No. H27445.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2497-21-4 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-21-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00010631
InChI Key FEWIGMWODIRUJM-HYXAFXHYSA-N
Synonym 4-hexen-3-one, 2-hexen-4-one, 2-hexene-4-one, e-hex-4-en-3-one, 4-hexene-3-one, fema no. 3352, e-4-hexen-3-one, 4-hexen-3-one, predominantly trans, e-ch3ch=chc =o c2h5, trans-4-hexen-3-one
PubChem CID 5365811
IUPAC Name (4Z)-hex-4-en-3-one
SMILES CCC(=O)\C=C/C

Specifications

Density 0.858
Boiling Point 135°C to 137°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.44
Quantity 10 g
UN Number UN1224
Formula Weight 98.15
Percent Purity 98%
Assay trans-isomer >95%
Chemical Name or Material 4-Hexen-3-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.