missing translation for 'onlineSavingsMsg'
Learn More

4-Hexen-1-ol, predominantly trans, 97%, Thermo Scientific Chemicals

Product Code. 15400758
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400758 5 g 5g
15410758 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400758 Supplier Thermo Scientific Alfa Aesar Supplier No. H54459.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Hexen-1-ol on reaction with NaIO4/NaHSO3 reagent yields 2-(1-iodoethyl)tetrahydrofuran. has been employed in the preparation of hex-4-enals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexen-1-ol on reaction with NaIO4/NaHSO3 reagent yields 2-(1-iodoethyl)tetrahydrofuran. has been employed in the preparation of hex-4-enals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-92-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009713
InChI Key VTIODUHBZHNXFP-NSCUHMNNSA-N
Synonym 4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans
PubChem CID 641248
IUPAC Name (E)-hex-4-en-1-ol
SMILES C\C=C\CCCO

Specifications

Density 0.851
Boiling Point 159°C to 160°C
Flash Point 61°C (142°F)
Linear Formula CH3CH=CH(CH2)3OH
Refractive Index 1.439
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material 4-Hexen-1-ol, predominantly trans
compliance-icons
Product Identifier
  • 4-Hexen-1-ol, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hexen-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.