missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanone, TRC

Product Code. 30395539
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395539 5 g -
30405440 10 g 10g
30400626 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395539 Supplier Toronto Research Chemicals Supplier No. TRCH2528305G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-19-3
Molecular Formula C7 H14 O
Molecular Weight (g/mol) 114.19
Synonym Heptan-4-one, Butyl propyl ketone
IUPAC Name heptan-4-one
SMILES CCCC(=O)CCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H14O/c1-3-5-7(8)6-4-2/h3-6H2,1-2H3
Formula Weight 114.1045
Chemical Name or Material 4-Heptanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.