missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanone, 98%

Product Code. 11364719
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
Unit Size:
1000mL
100mL
250mL
This item is not returnable. View return policy

Product Code. 11364719

Brand: Thermo Scientific Alfa Aesar B21222.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-19-3 | C7H14O | 114.188 g/mol

4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

Solubility
Miscible with alcohol, ligroin and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-19-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00009403
InChI Key HCFAJYNVAYBARA-UHFFFAOYSA-N
Synonym 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
PubChem CID 31246
IUPAC Name heptan-4-one
SMILES CCCC(=O)CCC

Specifications

Melting Point -33°C
Density 0.818
Boiling Point 143°C to 144°C
Flash Point 48°C (118°F)
Refractive Index 1.407
Quantity 100 mL
UN Number UN2710
Beilstein 1699049
Merck Index 14,3345
Solubility Information Miscible with alcohol,ligroin and ether. Immiscible with water.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 4-Heptanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.