missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanone, 98%

Product Code. 10785021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10785021 5 mL Glass bottle 5mL
10264810 500 mL Glass bottle 500mL
13478368 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10785021 Supplier Thermo Scientific Acros Supplier No. 146550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-19-3 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-19-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009403
InChI Key HCFAJYNVAYBARA-UHFFFAOYSA-N
Synonym 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
PubChem CID 31246
IUPAC Name heptan-4-one
SMILES CCCC(=O)CCC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -33.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 145.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2COCH2CH2CH3
Refractive Index 1.4060 to 1.4080
Quantity 5 mL
Beilstein 01,699
Merck Index 15,3386
Specific Gravity 0.81
Solubility Information Solubility in water: 4.6g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.73 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Heptanone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.