missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobutanoic acid, MedChemExpress

Product Code. 16435576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16435576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16435576 Supplier Medchem Express Supplier No. HY113286
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4-Guanidinobutanoic acid is a normal metabolite present in low concentrations.

4-Guanidinobutanoic acid is a normal metabolite present in low concentrations.

Specifications

Chemical Name or Material 4-Guanidinobutanoic acid
Physical Form Powder
CAS 463-00-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H11N3O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 25 mg/mL (172.22 mM; Need ultrasonic)
SMILES O=C(O)CCCNC(N)=N
Molecular Weight (g/mol) 145.16
Formula Weight 145.16
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.