missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic Acid Methanesulfonate, TRC

Product Code. 30397394
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397394 500 mg
30405522 100 mg
30405029 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397394 Supplier Toronto Research Chemicals Supplier No. TRCG368878500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148720-07-4
Molecular Formula C8H9N3O2 . CH4O3S
Molecular Weight (g/mol) 275.28
Synonym 4-Guanidinobenzoate Methanesulfonate
IUPAC Name 4-carbamimidamidobenzoic acid;methanesulfonic acid
SMILES CS(=O)(=O)O.NC(=N)Nc1ccc(cc1)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H9N3O2.CH4O3S/c9-8(10)11-6-3-1-5(2-4-6)7(12)13;1-5(2,3)4/h1-4H,(H,12,13)(H4,9,10,11);1H3,(H,2,3,4)
Formula Weight 275.0576
Chemical Name or Material 4-Guanidinobenzoic Acid Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.