missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenylboronic acid, 97%

Product Code. 10663563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10663563 5 g Glass bottle 5g
10467301 1 g Glass bottle 1g
10369273 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663563 Supplier Thermo Scientific Acros Supplier No. 359380050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87199-17-5 | C7H7BO3 | 149.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87199-17-5
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151823
InChI Key VXWBQOJISHAKKM-UHFFFAOYSA-N
Synonym 4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
PubChem CID 591073
IUPAC Name (4-formylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 256°C to 257°C
Color Colorless to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 149.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Formylphenylboronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Formylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: 4-Formylphenylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.