missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals

Product Code. 15427339
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427339 5 g 5g
15417339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427339 Supplier Thermo Scientific Alfa Aesar Supplier No. J66102.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the treatment of ischemic cardiovascular and cerebrovascular diseases. Also used in Chinese medicine in the treatment of headache and insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80154-34-3
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.264
MDL Number MFCD00210992
InChI Key OLZAGZCCJJBKNZ-SYLRKERUSA-N
Synonym helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
PubChem CID 12896796
IUPAC Name 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Solubility Information Soluble in hot water. Slightly soluble in water,ethanol or carbinol. Insoluble in aether or chloroform.
Optical Rotation −105° (c=0.8 in Water)
Formula Weight 284.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Formylphenyl beta-D-allopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.