missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenoxyacetic Acid 99%, ACROS Organics™

Product Code. 10641663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10641663 5 g Glass bottle 5g
10045760 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10641663 Supplier Thermo Scientific Acros Supplier No. 410780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22042-71-3
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00016613
InChI Key OYNIIKHNXNPSAG-UHFFFAOYSA-M
Synonym 4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid
PubChem CID 89177
IUPAC Name 2-(4-formylphenoxy)acetate
SMILES [O-]C(=O)COC1=CC=C(C=O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Formylphenoxyacetic acid, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.