missing translation for 'onlineSavingsMsg'
Learn More

4-Formylmorpholine, 99%

Product Code. p-7026953
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11430927

Brand: Thermo Scientific Alfa Aesar B23315.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4394-85-8 | C5H9NO2 | 115.132 g/mol

4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O

Specifications

Melting Point 20°C to 23°C
Density 1.15
pH 10
Boiling Point 239°C to 241°C
Flash Point 118°C (244°F)
Refractive Index 1.486
Quantity 100 g
Beilstein 110293
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material 4-Formylmorpholine
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.