missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzoic Acid Methyl Ester, TRC

Product Code. 30404806
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404806 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404806 Supplier Toronto Research Chemicals Supplier No. TRCF697410100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Formylbenzoic Acid Methyl Ester
CAS 1571-08-0
Molecular Formula C9H8O3
Quantity 100 g
Synonym Terephthalaldehydic Acid Methyl Ester, 4-(Methoxycarbonyl)benzaldehyde, 4-Carbomethoxybenzaldehyde, 4-Carboxybenzaldehyde Methyl Ester, 4-Formylbenzoic Acid Methyl Ester, Methyl 4-formylbenzoate, Methyl Benzaldehyde-4-carboxylate, Methyl p-Formylbenzoate, Methyl Terephthalaldehydate, NSC 28459, Para(methoxycarbonyl)benzaldehyde, p-(Methoxycarbonyl)benzaldehyde, p-Carbomethoxybenzaldehyde, p-Formylbenzoic Acid Methyl Ester
InChI Formula InChI=1S/C9H8O3/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-6H,1H3
SMILES COC(=O)c1ccc(C=O)cc1
IUPAC Name methyl 4-formylbenzoate
Molecular Weight (g/mol) 164.16
Formula Weight 164.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.